Learn More
Biomatik Corporation Human Glutamyl Prolyl tRNA Synthetase (EPRS) ELISA Kit
Supplier:  Biomatik Corporation EKU12453
Encompass_Preferred
Human Glutamyl Prolyl tRNA Synthetase (EPRS) ELISA Kit for detecting EPRS in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P07814. Gene ID: 2058. Target Synonyms: PARS; EARS; PIG32; QPRS; QARS; Bifunctional Aminoacyl-TRNA Synthetase; Glutamate tRNA Ligase; Proline tRNA Ligase; Cell proliferation-inducing gene 32 protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7037