Learn More
Biomatik Corporation Human Glutaredoxin 2 (GLRX2) ELISA Kit
Supplier:  Biomatik Corporation EKU11788
Encompass_Preferred
Human Glutaredoxin 2 (GLRX2) ELISA Kit for detecting GLRX2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.054ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9NS18. Gene ID: 51022. Target Synonyms: GRX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6777