Learn More
Abcam Human Glutathione Peroxidase 4 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502660583
Encompass_Preferred
Human Glutathione Peroxidase 4 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Glutathione Peroxidase 4.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-0583 Supplier Abcam Supplier No. AB30925810X96TESTS
Human Glutathione Peroxidase 4 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human Glutathione Peroxidase 4.
The product is subject to the following: Abcam Restricted Use Statement