Learn More
American Research Products Inc Human Glutathione S transferas
Catalog No. 5000327702
Encompass
Human Glutathione S-transferase omega-1 (GSTO1) ELISA kit from CUSABIO Detection Range 31 2 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich
Catalog No. 50-003-27702 Supplier American Research Products Inc Supplier No. CSBEL009986HU
Human Glutathione S-transferase omega-1 (GSTO1) ELISA kit from CUSABIO Detection Range 31 2 pg/mL-2000 pg/mL Sensitivity 7 8 pg/mL Sample serum plasma tissue homogenates cell lysates Method Sandwich