Learn More
Biomatik Corporation Human Glutathione Synthetase (GSS) ELISA Kit
Supplier:  Biomatik Corporation EKU04500
Encompass_Preferred
Human Glutathione Synthetase (GSS) ELISA Kit for detecting GSS in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P48637. Gene ID: 2937. Target Synonyms: Glutathione synthase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5902