Learn More
American Research Products Inc Human Glycated Albumin GA ELI
Catalog No. 5000329335
Encompass
Human Glycated Albumin GA ELISA Kit from CUSABIO Detection Range 18 75 pmol/mL-1200 pmol/mL Sensitivity 4 7 pmol/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29335 Supplier American Research Products Inc Supplier No. CSBE09599H
Human Glycated Albumin GA ELISA Kit from CUSABIO Detection Range 18 75 pmol/mL-1200 pmol/mL Sensitivity 4 7 pmol/mL Sample serum plasma Method Competitive