Learn More
Biomatik Corporation Human Glycated Hemoglobin A1c (HbA1c) CLIA Kit
Supplier:  Biomatik Corporation EKU10875
Encompass_Preferred
Human Glycated Hemoglobin A1c (HbA1c) CLIA Kit for detecting HbA1c in serum, plasma, erythrocyte lysates. Species Reactivity: Human (Homo sapiens). Detection Range: 5.86-1,500ug/ml. Sensitivity: 2.44ug/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: O95602. Gene ID: 25885. Target Synonyms: Glycosylated Hemoglobin; Hemoglobin A1c; Hb1c; HbAIc; HbAIc. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5435