Learn More
Biomatik Corporation Human Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) CLIA Kit
Supplier:  Biomatik Corporation EKU08949
Encompass_Preferred
Human Glyceraldehyde-3-Phosphate Dehydrogenase (GAPDH) CLIA Kit for detecting GAPDH in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.27-200ng/ml. Sensitivity: 0.12ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P04406. Gene ID: 2597. Target Synonyms: G3PD; GAPD; Peptidyl-cysteine S-nitrosylase GAPDH. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-7053