Learn More
Biomatik Corporation Human Glycine Dehydrogenase (GLDC) ELISA Kit
Supplier:  Biomatik Corporation EKU04508
Encompass_Preferred
Human Glycine Dehydrogenase (GLDC) ELISA Kit for detecting GLDC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.132ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P23378. Gene ID: 2731. Target Synonyms: GCSP; NKH; Decarboxylating; Glycine Decarboxylase; Glycine Cleavage System Protein P; Glycine dehydrogenase (aminomethyl-transferring). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8952