Learn More
Biomatik Corporation Human Glycine Dehydrogenase (GLDC) CLIA Kit
Supplier:  Biomatik Corporation EKU09424
Encompass_Preferred
Human Glycine Dehydrogenase (GLDC) CLIA Kit for detecting GLDC in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 13.7-10,000pg/ml. Sensitivity: 4.7pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P23378. Gene ID: 2731. Target Synonyms: GCSP; NKH; Decarboxylating; Glycine Decarboxylase; Glycine Cleavage System Protein P; Glycine dehydrogenase (aminomethyl-transferring). Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6395