Learn More
Bioworld HUMAN GLYCOGEN PHOSPHORYLAS
Supplier:  Bioworld 220603361
Encompass_Preferred
HUMAN GLYCOGEN PHOSPHORYLASE (RECOMBINANT): Recombinant Human Glycogen Phosphorylase (GPBB) Source:E.Coli Background: Glycogen Phosphorylase is an enzyme that catalyzes the degradation of Glycogen in animals by releasing glucose-1-pho...
Catalog No. 50-253-433