Learn More
Biomatik Corporation Human Glycogen Phosphorylase, Muscle (PYGM) ELISA Kit
Supplier:  Biomatik Corporation EKU04518
Encompass_Preferred
Human Glycogen Phosphorylase, Muscle (PYGM) ELISA Kit for detecting PYGM in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.312-20ng/ml. Sensitivity: 0.115ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P11217. Gene ID: 5837. Target Synonyms: GPMM; Phosphorylase,Glycogen; McArdle Syndrome,Glycogen Storage Disease Type V; Myophosphorylase. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8949