Learn More
Biomatik Corporation Human Glycogen Synthase Kinase 3 Alpha (GSK3a) ELISA Kit
Supplier:  Biomatik Corporation EKU04520
Encompass_Preferred
Human Glycogen Synthase Kinase 3 Alpha (GSK3a) ELISA Kit for detecting GSK3a in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.049ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49840. Gene ID: 2931. Target Synonyms: Serine/threonine-protein kinase GSK3A. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4336