Learn More
American Research Products Inc Human glycoprotein 130 gp130
Catalog No. 5000326091
Encompass
Human glycoprotein 130 gp130 ELISA KIT from CUSABIO Detection Range 0 938 ng/mL-60 ng/mL Sensitivity 0 516 ng/mL Sample serum plasma tissue homogenates saliva urine Method Sandwich
Catalog No. 50-003-26091 Supplier American Research Products Inc Supplier No. CSBE04571H
Human glycoprotein 130 gp130 ELISA KIT from CUSABIO Detection Range 0 938 ng/mL-60 ng/mL Sensitivity 0 516 ng/mL Sample serum plasma tissue homogenates saliva urine Method Sandwich