Learn More
Biomatik Corporation Human Glycosylphosphatidylinositol Anchored High Density Lipoprotein Binding Protein 1 (GPIHBP1) ELISA Kit
Supplier:  Biomatik Corporation EKU04535
Encompass_Preferred
Human Glycosylphosphatidylinositol Anchored High Density Lipoprotein Binding Protein 1 (GPIHBP1) ELISA Kit for detecting GPIHBP1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.625-10ng/ml. Sensitivity: 0.242ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IV16. Gene ID: 338328. Target Synonyms: GPI-HBP1; High density lipoprotein-binding protein 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5422