Learn More
Biomatik Corporation Human Glycosylphosphatidylinositol Anchored Molecule Like Protein (GML) ELISA Kit
Supplier:  Biomatik Corporation EKU04536
Encompass_Preferred
Human Glycosylphosphatidylinositol Anchored Molecule Like Protein (GML) ELISA Kit for detecting GML in tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99445. Gene ID: 2765. Target Synonyms: LY6DL; GPI Anchored Molecule Like Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4426