Learn More
Biomatik Corporation Human Glycosyltransferase 25 Domain Containing Protein 2 (GLT25D2) ELISA Kit
Supplier:  Biomatik Corporation EKU10879
Encompass_Preferred
Human Glycosyltransferase 25 Domain Containing Protein 2 (GLT25D2) ELISA Kit for detecting GLT25D2 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.055ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8IYK4. Gene ID: 23127. Target Synonyms: C1orf17; Procollagen galactosyltransferase 2; Collagen beta (1-O) galactosyltransferase 2; Hydroxylysine galactosyltransferase 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6568