Learn More
Raybiotech Inc Human Glypican 1 ELISA Kit
Supplier:  Raybiotech Inc ELHGPC11
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 0.15 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes Human Glypican-1 (GPC1)
Catalog No. 50-194-9325