Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Glypican 1 ELISA Kit

Catalog No. EH222RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH222RB 96 Tests
EH222RBX5 5 x 96 Tests
EH222RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH222RB Supplier Invitrogen™ Supplier No. EH222RB
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ELISA

Human Glypican 1 quantitates human Glypican 1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human Glypican 1.

Cell surface heparan sulfate proteoglycans are composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P35052
Assay Range 0.15-40 ng/mL
Assay Sensitivity 0.15 ng/mL
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias AI462976; glypican; glypican 1; glypican proteoglycan 1; glypican-1; Gpc1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; HSPGM12; Secreted glypican-1AI462976; glypican; glypican 1; glypican proteoglycan 1; glypican-1; Gpc1; GPI-anchored cell surface heparan sulfate proteoglycan; HSPG M12; HSPGM12; Secreted glypican-1;
Gene ID (Entrez) 2817
Gene Symbol GPC1
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Shipping Condition Wet or Dry Ice
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Target Glypican 1
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.