Learn More
ACROBiosystems Human Glypican 3 (S495A, S509A), His Tag (GP3-H52H8) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1 (S495A, S509A)).
Catalog No. 502253081
Encompass_Preferred
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.
This item is not returnable. View return policy
Catalog No. 50-225-3081 Supplier ACROBiosystems Supplier No. GP3H52H8100UG
Glypican-3 (GPC3) is also known as Intestinal protein OCI-5, GTR2-2, MXR7, which belongs to the glypican family. Glypican 3 / GPC-3 is highly expressed in lung, liver and kidney. Glypican-3 inhibits the dipeptidyl peptidase activity of DPP4. Glypican-3 may be involved in the suppression/modulation of growth in the predominantly mesodermal tissues and organs, and also may play a role in the modulation of IGF2 interactions with its receptor and thereby modulate its function.