Learn More
ACROBiosystems APC-Labeled Human Glypican 3 / GPC3 Protein, His Tag (Site-specific conjugation)
Catalog No. 502331845
Encompass_Preferred
APC-Labeled Human Glypican 3, His Tag (GP3-HA2H9) is produced via conjugation of APC to Human Glypican 3, His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human Glypican 3, His Tag is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).
Catalog No. 50-233-1845 Supplier ACROBiosystems Supplier No. GP3HA2H925TESTS
APC-Labeled Human Glypican 3, His Tag (GP3-HA2H9) is produced via conjugation of APC to Human Glypican 3, His Tag with a new generation site-specific technology under optimal conditions with a proprietary technology. Human Glypican 3, His Tag is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1).