Learn More
Raybiotech Inc Human Glypican 3 ELISA Kit
Supplier:  Raybiotech Inc ELHGPC31
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.8 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Glypican 3. Other species not determined.
Catalog No. 50-194-9326