Learn More
ACROBiosystems ACROBiosystems Human Glypican 3 / GPC3 (S359F) Protein, His Tag, low endotoxin
Catalog No. 502106651
Encompass_Preferred
Recombinant Protein;1MG;Human Glypican 3 (S359F), His Tag, low endotoxin (GP3-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1 (S359F)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.7 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 0.5 μg/mL (100 μL/well) can bind Human Glypican 3 (S359F), His Tag, low endotoxin (Cat. No. GP3-H5223) with a linear range of 3-128 ng/mL (QC tested).;Immobilized Human Glypican 3 (S359F), His Tag, low endotoxin (Cat. No. GP3-H5223) at 0.5 μg/mL (100 μL/well) can bind Anti-Human GPC3 Mab with a linear range of 0.4-3 ng/mL (Routinely tested).;Anti-Human GPC3 MAb (chimeric mouse-human IgG1) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human Glypican 3 (S359F), His Tag, low endotoxin (Cat. No. GP3-H5223) with an a
Catalog No. 50-210-6651 Supplier ACROBiosystems Supplier No. GP3H52231MG
Recombinant Protein;1MG;Human Glypican 3 (S359F), His Tag, low endotoxin (GP3-H5223) is expressed from human 293 cells (HEK293). It contains AA Gln 25 - His 559 (Accession # P51654-1 (S359F)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 61.7 kDa. The protein migrates as 66-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGF basic, Tag Free (Cat. No. BFF-H4117) at 0.5 μg/mL (100 μL/well) can bind Human Glypican 3 (S359F), His Tag, low endotoxin (Cat. No. GP3-H5223) with a linear range of 3-128 ng/mL (QC tested).;Immobilized Human Glypican 3 (S359F), His Tag, low endotoxin (Cat. No. GP3-H5223) at 0.5 μg/mL (100 μL/well) can bind Anti-Human GPC3 Mab with a linear range of 0.4-3 ng/mL (Routinely tested).;Anti-Human GPC3 MAb (chimeric mouse-human IgG1) captured on CM5 chip via anti-human IgG Fc antibodies surface, can bind Human Glypican 3 (S359F), His Tag, low endotoxin (Cat. No. GP3-H5223) with an a