Learn More
Raybiotech Inc Human GM-CSF R alpha ELISA Kit
Supplier:  Raybiotech Inc ELHGMCSFRA1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.3 ng/ml.. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human GM-CSF R alpha.
Catalog No. 50-194-9322