Learn More
Sino Biological Anti-GNGT1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13658RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GNGT1 / GNG1 (rh GNGT1 / GNG1; Catalog#13658-H07E; P63211; Pro2-Cys71). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-240-8971