Learn More
BIOHIPPO Human GNLY Microsample
SDP
Catalog No. 5000314525
Encompass
Human GNLY Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GNLY in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 24-15 ng/mL with a sensitivity of 0 08 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GNLY Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GNLY Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GNLY biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14525 Supplier BIOHIPPO Supplier No. BHE15212135
Human GNLY Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GNLY in serum plasma tissue homogenates cell lysates cell culture supernates and other biological fluids It provides a detection range of 0 24-15 ng/mL with a sensitivity of 0 08 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GNLY Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GNLY Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GNLY biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color