Learn More
Sino Biological Human GNLY Protein 500ug (Sino Biological)
Supplier:  Sino Biological 10320H02H500
Encompass_Preferred
A DNA sequence encoding the human GNLY (P22749-1) (Arg23-Leu145) was expressed with the fused Fc region of human IgG1 at the C-terminus.
Catalog No. 50-237-3231