Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human GnRH1 Synthetic Peptide

Catalog No. PIPEP10933 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPEP10933 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPEP10933 Supplier Invitrogen™ Supplier No. PEP10933
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic Peptide

PEP-10933 is a synthetic peptide suitable for use as a control in ELISA and Western blot applications. PEP-10933 has the amino acid sequence: Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2. Store lyophilized product at -20 C. Upon reconstitution store at 4C for 2-7 days or aliquot and store at -20C. For long term storage add HSA or BSA to 0.1%. Avoid repeated freeze/thaw cycles. Reconstitute the lyophilized product in sterile water not less than 100 ug/ml.

The Luteinizing Hormone Releasing Hormone (LH-RH) is produced in the hypothalamus and influences the release of Luteinizing Hormone in the pituitary gland. LH-RH is also known as Gonadotropin-releasing hormone (Gn-RH).
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 2796
Species Human
Purification Method purified
Accession Number P01148
Common Name GnRH1
Content And Storage -20°C. Avoid freeze-thaw cycles.
Description Synthetic Peptide
Format Lyophilized
Formulation Peptide with no preservative
For Use With (Application) Control
Gene Alias Gnrh; Gnrh1; Gnrh2; Gnrha; GnRH-associated peptide 1; GnRH-associated peptide I; GnRH-I; Gonadoliberin I; Gonadoliberin-1; Gonadorelin; gonadotropin releasing hormone 1; gonadotropin releasing hormone 2; gonadotropin-releasing hormone 1 (luteinizing-releasing hormone); gonadotropin-releasing hormone 1 variant A; gonadotropin-releasing hormone 1 variant B; Gonadotropin-releasing hormone I; GRH; hpg; hypogonadal; leuteinizing-releasing hormone; LHRH; LH-RH I; Lhrh1; LNRH; Luliberin I; luteinizing hormone releasing hormone; luteinizing hormone-releasing hormone; luteinizing hormone-releasing hormone I; Lutropin; Progonadoliberin I; progonadoliberin-1; prolactin release-inhibiting factor; Prolactin release-inhibiting factor 1; Prolactin release-inhibiting factor I; Rgnrhg1
Gene Symbol GNRH1
Lyophilized Yes
Preservative No preservative
Product Type Peptide
Quantity 10 mg
Regulatory Status RUO
Sequence Pyr-His-Trp-Ser-Tyr-Gly-Leu-Arg-Pro-Gly-NH2
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.