Learn More
Biomatik Corporation Human Golgi Glycoprotein 1 (GLG1) ELISA Kit
Supplier:  Biomatik Corporation EKU12248
Encompass_Preferred
Human Golgi Glycoprotein 1 (GLG1) ELISA Kit for detecting GLG1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 37pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q92896. Gene ID: 2734. Target Synonyms: CFR-1; ESL-1; MG160; Golgi Apparatus Protein 1; E-selectin ligand 1; Cysteine-rich fibroblast growth factor receptor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4758