Learn More
Sino Biological Anti-PIST Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 14911T26100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PIST (rh PIST; Catalog#14911-H07E; Q9HD26-1; Ile286-Tyr462). PIST specific IgG was purified by Human PIST affinity chromatography.
Catalog No. 50-255-4118