Learn More
Sino Biological Human GOT1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 14196T60100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Aspartate aminotransferase / GOT1 (rh Aspartate aminotransferase / GOT1; Catalog#14196-H07E; P17174; Met1-Gln413). Aspartate aminotransferase / GOT1 specific IgG was purified by Human Aspartate aminotransferase / GOT1 affinity chromatography.
Catalog No. 50-237-3525