Learn More
Sino Biological Recombinant Human CD42b/GP1BA Protein (His Tag), Amine-Labeled Biotinylated
Catalog No. 502993517
Encompass_Preferred
A DNA sequence encoding the Human GP1BA (CAM97225.1) (Met1-Gly519) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-299-3517 Supplier Sino Biological Supplier No. 11765H08HB100
A DNA sequence encoding the Human GP1BA (CAM97225.1) (Met1-Gly519) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.