Learn More
BIOHIPPO Human GP5 Microsample
SDP
Catalog No. 5000313885
Encompass
Human GP5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GP5 in Plasma It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 122 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GP5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GP5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GP5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13885 Supplier BIOHIPPO Supplier No. BHE15211658
Human GP5 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GP5 in Plasma It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 122 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GP5 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GP5 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GP5 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color