Learn More
ACROBiosystems ACROBiosystems Human GPA33 / A33 Protein, Fc Tag
Supplier:  ACROBiosystems GP3H525350UG
Encompass_Preferred
Recombinant Protein;50UG;Human GPA33, Fc Tag (GP3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # Q99795-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50 kDa.
Catalog No. 50-210-6273