Learn More
ACROBiosystems ACROBiosystems Human GPA33 / A33 Protein, Fc Tag
Catalog No. 502106274
Encompass_Preferred
Recombinant Protein;1MG;Human GPA33, Fc Tag (GP3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # Q99795-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50 kDa.
Catalog No. 50-210-6274 Supplier ACROBiosystems Supplier No. GP3H52531MG
Recombinant Protein;1MG;Human GPA33, Fc Tag (GP3-H5253) is expressed from human 293 cells (HEK293). It contains AA Ile 22 - Val 235 (Accession # Q99795-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 50 kDa.