Learn More
Sino Biological Human GPA33 / Glycoprotein-A33 Protein (His Tag) 500ug
Supplier:  Sino Biological 11277H08H500
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human GPA33 (Q99795-1) (Met 1-Val 235) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-224-6879