Learn More
Sino Biological Recombinant Anti-GPA33/A33 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 11277R07120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human GPA33 (rh GPA33; Catalog#11277-H08H; Q99795-1; Met1-Val235).
Catalog No. 50-255-4746