Learn More
Sino Biological Recombinant Human Glypican 3/GPC3 Protein (495S/A, mFc Tag)
Supplier:  Sino Biological 10088H38H50
Encompass_Preferred
A DNA sequence encoding the human GPC3 (NP_004475.1, with mutation Ser 495 Ala) (Met1-Ser550) was expressed with the Fc region of mouse IgG2a at the C-terminus.
Catalog No. 50-255-5484