Learn More
Sino Biological Anti-GPIHBP1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 15388RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human GPIHBP1 / GPI-HBP1 (rh GPIHBP1 / GPI-HBP1; Catalog#15388-H08H; AAH63857.1; Met1-Ser160). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4107