Learn More
Sino Biological Anti-Osteoactivin/GPNMB Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 201527T02100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human GPNMB fragment, and purified by antigen affinity chromatography.
Catalog No. 50-241-2498