Learn More
BIOHIPPO Human GPR172A Microsample
SDP
Catalog No. 5000314681
Encompass
Human GPR172A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPR172A in Tissue homogenates saliva and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 61 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GPR172A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GPR172A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GPR172A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14681 Supplier BIOHIPPO Supplier No. BHE15211383
Human GPR172A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPR172A in Tissue homogenates saliva and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 61 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GPR172A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GPR172A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GPR172A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color