Learn More
BIOHIPPO Human GPR39 Microsample
SDP
Catalog No. 5000313856
Encompass
Human GPR39 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPR39 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 107 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GPR39 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GPR39 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GPR39 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13856 Supplier BIOHIPPO Supplier No. BHE15210597
Add to Cart
Add to Cart
Human GPR39 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPR39 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 107 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GPR39 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GPR39 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GPR39 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color