Learn More
BIOHIPPO Human GPRC5A Microsample
SDP
Catalog No. 5000314471
Encompass
Human GPRC5A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPRC5A in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 116 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GPRC5A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GPRC5A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GPRC5A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14471 Supplier BIOHIPPO Supplier No. BHE15211066
Human GPRC5A Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of GPRC5A in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 116 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human GPRC5A Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human GPRC5A Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human GPRC5A biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color