Learn More
ACROBiosystems Biotinylated Human GPRC5D Protein, His,Avitag™&Flag Tag (Nanodisc)
Supplier:  ACROBiosystems GPDH82D4100UG
Encompass_Preferred
Biotinylated Human GPRC5D, His,Avitag&Flag Tag (Nanodisc) (GPD-H82D4) is expressed from human 293 cells (HEK293). It contains AA Tyr 2 - Phe 192 & Glu 197 - Leu 262 (Accession # Q9NZD1-2).
Catalog No. 50-233-1662