Learn More
Sino Biological Anti-GPRC5D Antibody, Mouse Monoclonal
Supplier:  Sino Biological 24447MM12100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPRC5D. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-3851