Learn More
Sino Biological Anti-GPRC5D Antibody (FITC), Mouse Monoclonal
Supplier:  Sino Biological 24447MM19F25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human GPRC5D. The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography, and conjugated with FITC under optimum conditions, the unreacted FITC was removed.
Catalog No. 50-255-3840