Learn More
Sino Biological Human GPRC5D Protein 20ug (Sino Biological)
Supplier:  Sino Biological 24447H18HDD20
Encompass_Preferred
A DNA sequence encoding the human GPRC5D (Q9NZD1) (Met1-Ser268) was expressed, with a Flag tag at the N-terminus and a polyhistidine tag at the C-terminus.
Catalog No. 50-237-3881