Learn More
Bon Opus Biosciences Recombinant Human GPX4 (C-His) 50ug
SDP
Supplier:  Bon Opus Biosciences BP109750UG
Encompass_Preferred
Recombinant Human GPX4 (C-His) is produced by Human Cells. The target gene encoding Met1-Phe197 is expressed with a C-His tag. Accession# P36969. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, pH 7.4. Bon Opus Cat# BP1097
Catalog No. 50-217-2695