Learn More
Sino Biological Human GPX4 Protein 100ug (Sino Biological)
Supplier:  Sino Biological 29993H07E100
Encompass_Preferred
A DNA sequence encoding the Human GPX4 (P36969) (Gly26-Phe197,U73C) was expressed, with a polyhistidine tag at the N-terminus.
Catalog No. 50-237-2362