Learn More
Biomatik Corporation Human Granulin (GRN) CLIA Kit
Supplier:  Biomatik Corporation EKU09819
Encompass_Preferred
Human Granulin (GRN) CLIA Kit for detecting GRN in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.14-100ng/ml. Sensitivity: 0.07ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P28799. Gene ID: 2896. Target Synonyms: GEP; GP88; PCDGF; PEPI; PGRN; Proepithelin; Acrogranin; Glycoprotein of 88 Kda; Paragranulin. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8428